Assay Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001492.2 | NP_001001492.2 | 3 - 4 | 307 | 68 | |
| GenBank mRNA | AK029554.1 | - | 3 - 4 | 301 | 68 | |
| AK133316.1 | - | 3 - 4 | 307 | 68 | ||
| BC141285.1 | - | 2 - 3 | 157 | 9055897 | 68 | |
| BC145611.1 | - | 2 - 3 | 157 | 9054177 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029554.1, BC141285.1, BC145611.1, NM_001001492.2 transcripts.
Set Membership: |
|

Lca5l
Genomic Map