Assay Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001492.2 | NP_001001492.2 | 6 - 7 | 1292 | 136 | |
| GenBank mRNA | AK029554.1 | - | 6 - 7 | 1286 | 136 | |
| AK133316.1 | - | 6 - 7 | 1293 | 136 | ||
| BC141285.1 | - | 5 - 6 | 1142 | 9055897 | 136 | |
| BC145611.1 | - | 5 - 6 | 1142 | 9054177 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lca5l
Genomic Map