Assay Details
| Gene Symbol: Mtus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029920.7 | NP_084196.4 | 11 - 12 | 4464 | 151 | |
| GenBank mRNA | AB093263.1 | - | 10 - 11 | 3338 | 151 | |
| AK021162.1 | - | 5 - 6 | 666 | 151 | ||
| AK147457.1 | - | 11 - 12 | 4465 | 151 | ||
| BC055016.1 | - | 5 - 6 | 707 | 151 | ||
| BC085194.1 | - | 11 - 12 | 3810 | 151 | ||
More Information
Set Membership: |
|

Mtus2
Genomic Map