Assay Details
| Gene Symbol: 4930500F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045758.1 | 2 - 3 | 280 | 75 | ||
| GenBank mRNA | AK015658.1 | - | 2 - 3 | 280 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 21221 |
Gm21221 | -36.7kb | Anti-sense | Chr. 5 - 149801990 - 149811751 on NCBI Build GRCm38 | |
| Downstream | relaxin/insulin-like family peptide receptor 2 | Rxfp2 |
Gpr106, Great, Lgr8 |
164.9kb | Sense | Chr. 5 - 150017781 - 150082550 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930500F04Rik
Genomic Map