Assay Details
| Gene Symbol: Gm21221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077242.1 | 3 - 4 | 554 | 67 | ||
| GenBank mRNA | AK076945.1 | - | 3 - 4 | 557 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | beta-3-glucosyltransferase | B3glct |
B3galtl, Gm1057 |
-40.9kb | Anti-sense | Chr. 5 - 149678105 - 149762599 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930500F04 gene | 4930500F04Rik | 4931407C17Rik | 43.9kb | Anti-sense | Chr. 5 - 149847747 - 149864280 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm21221
Genomic Map