| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1420214_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1420216_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1447595_PM_x_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1420215_PM_x_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1447594_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447595_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447595_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420214_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420215_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447594_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420215_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447594_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420216_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420214_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420216_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0800002070.mm.1 |
NR_045473.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002070.mm.1 |
NR_045473.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002070.mm.1 |
TC0800000275.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1447595_x_at |
- |
- |
| Mouse Expression Set 430 |
1420215_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1420214_PM_at |
- |
- |
| Mouse Expression Set 430 |
1420214_at |
- |
- |
| Mouse Expression Set 430 |
1447594_at |
- |
- |
| Mouse Expression Set 430 |
1420216_at |
- |
- |
| Mouse Expression Set 430 |
1420215_x_at |
- |
- |
| Mouse Expression Set 430 |
1447595_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1420216_PM_at |
- |
- |
| Mouse Expression Set 430 |
1447594_PM_at |
- |
- |
| Murine Genome U74v2 Set |
167206_PM_at |
- |
- |
| Murine Genome U74v2 Set |
167206_at |
- |
- |
| Murine Genome U74v2 Set |
167205_at |
- |
- |
| Murine Genome U74v2 Set |
167205_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1447594_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1447595_x_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1447594_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1447595_PM_x_at |
- |
- |