Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36017 |
Gm36017 | -213.8kb | Anti-sense | Chr. 5 - 44412732 - 44427652 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930431F12 gene | 4930431F12Rik | 314.4kb | Sense | Chr. 5 - 44966201 - 44978805 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map