Assay Details
Gene Symbol: Ppp2r2cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045505.1 | 2 - 3 | 1310 | 91 | ||
GenBank mRNA | AK016607.1 | - | 2 - 3 | 1306 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
protein phosphatase 2, regulatory subunit B, gamma |
Ppp2r2c |
6330548O06Rik, IMYPNO, IMYPNO1, PR52 |
Anti-sense | Chr. 5 - 36866843 - 36955078 on NCBI Build GRCm38 | |
Downstream | Wolfram syndrome 1 homolog (human) | Wfs1 |
AI481085, wolframin |
91.1kb | Sense | Chr. 5 - 36966104 - 36988982 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
