Assay Details
| Gene Symbol: Cpsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164272.1 | NP_001157744.1 | 1 - 2 | 91 | 52 | |
| NM_172302.3 | NP_758506.3 | 1 - 2 | 91 | 52 | ||
| GenBank mRNA | AK044306.1 | - | 1 - 2 | 70 | 52 | |
| AK053375.1 | - | 1 - 2 | 70 | 52 | ||
| AK088603.1 | - | 1 - 2 | 69 | 52 | ||
| AK165330.1 | - | 1 - 2 | 69 | 52 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK053375.1, AK088603.1, AK165330.1, NM_001164272.1, NM_172302.3 transcripts.
Set Membership: |
|

Cpsf7
Genomic Map