Assay Details
| Gene Symbol: Cpsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164272.1 | NP_001157744.1 | 6 - 7 | 1076 | 118 | |
| NM_172302.3 | NP_758506.3 | 6 - 7 | 1103 | 118 | ||
| GenBank mRNA | AK044306.1 | - | 6 - 7 | 1083 | 118 | |
| AK053375.1 | - | 7 - 8 | 1148 | 118 | ||
| AK088603.1 | - | 6 - 7 | 1081 | 118 | ||
| AK165330.1 | - | 6 - 7 | 1081 | 118 | ||
| BC017519.1 | - | 5 - 6 | 996 | 118 | ||
| BC038812.1 | - | 5 - 6 | 1006 | 4506179 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf7
Genomic Map