Assay Details
| Gene Symbol: Aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276284.1 | NP_001263213.1 | 5 - 6 | 928 | 97 | |
| NM_016666.3 | NP_057875.1 | 6 - 7 | 1027 | 97 | ||
| GenBank mRNA | AK011609.1 | - | 5 - 6 | 801 | 97 | |
| AK146390.1 | - | 5 - 6 | 930 | 97 | ||
| BC075614.1 | - | 5 - 6 | 799 | 5710504 | 97 | |
| BQ175443.1 | - | 1 - 2 | 97 | |||
| U85489.1 | - | 6 - 7 | 996 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
protein targeting to mitochondrion
xenobiotic metabolic process
regulation of protein kinase A signaling
protein maturation by protein folding
negative regulation of cyclic-nucleotide phosphodiesterase activity
regulation of nucleic acid-templated transcription
transcription cofactor activity
protein binding
aryl hydrocarbon receptor binding
GAF domain binding
unfolded protein binding
protein folding
protein targeting to mitochondrion
xenobiotic metabolic process
regulation of protein kinase A signaling
protein maturation by protein folding
negative regulation of cyclic-nucleotide phosphodiesterase activity
regulation of nucleic acid-templated transcription
transcription cofactor activity
protein binding
aryl hydrocarbon receptor binding
GAF domain binding
unfolded protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921203 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921208 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921206 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115704 | BC075614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115704 | U85489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115704 | NM_016666.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115704 | AK011609.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115704 | AK146390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125173 | BC075614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125173 | U85489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125173 | NM_016666.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125173 | AK011609.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125173 | AK146390.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508976 | BC089593.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508976 | NM_153069.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130220 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125173 | BC075614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125173 | U85489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125173 | NM_016666.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125173 | AK011609.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125173 | AK146390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115704 | BC075614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115704 | U85489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115704 | NM_016666.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115704 | AK011609.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115704 | AK146390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205737 | BC075614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205737 | U85489.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205737 | NM_016666.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205737 | AK011609.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205737 | AK146390.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921225 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09089724 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09089749 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03540627 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03540630 | - | - |
| Whole Mouse Genome G4122A | A_51_P205737 | BC075614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205737 | U85489.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205737 | NM_016666.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205737 | AK011609.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205737 | AK146390.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119866 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125173 | BC075614.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125173 | U85489.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125173 | NM_016666.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125173 | AK011609.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125173 | AK146390.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09089724 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14312749 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14206900 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14377077 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921203 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921206 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921208 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31921226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14465006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14152237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14389356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14396092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14417695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14396096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14199538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14295382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14264778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14152088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14336561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14152090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14152235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14499838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14377077 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10464607 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464608 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464611 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423128_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10464603 | - | - |
| Mouse Genome 430 2.0 Array | 1423128_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423128_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11632_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11632_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900007815.mm.1 | NM_001276284.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007815.mm.1 | NM_016666.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007815.mm.1 | TC1900000903.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900007818.mm.1 | NM_001276284.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007818.mm.1 | NM_016666.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007818.mm.1 | TC1900000903.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423128_at | - | - |
| Mouse Expression Set 430 | 1423128_PM_at | - | - |
| Murine Genome U74v2 Set | 92575_PM_at | - | - |
| Murine Genome U74v2 Set | 92575_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423128_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423128_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423128_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423128_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 430511 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5720300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720300 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777655 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777655 | - | - |
| mouse-6 v1.1 (Illumina) | scl53441.6.1_144-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53441.6.1_144-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2777655 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777655 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53441.6.1_144-S | - | - |

Aip
Genomic Map