Assay Details
| Gene Symbol: 4930404I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028368.1 | 1 - 2 | 70 | 65 | ||
| GenBank mRNA | AK006191.1 | - | 1 - 2 | 61 | 65 | |
| AK015081.1 | - | 1 - 2 | 72 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

4930404I05Rik
Genomic Map