| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600016156.mm.1 |
NR_040486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016156.mm.1 |
NR_040486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016156.mm.1 |
TC1600002103.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040489.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040489.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040490.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040490.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040491.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
NR_040491.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016154.mm.1 |
TC1600002103.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
NR_040486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
NR_040486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
NR_040487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
NR_040487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
NR_040491.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
NR_040491.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600016155.mm.1 |
TC1600002103.mm.1 |
MTA TC |