Assay Details
| Gene Symbol: Gm11832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050144.1 | - | 1 - 2 | 64 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibrinogen silencer binding protein | Fsbp | -24.4kb | Anti-sense | Chr. 4 - 11579662 - 11587802 on NCBI Build GRCm38 | |
| Downstream | GTP binding protein (gene overexpressed in skeletal muscle) | Gem | AV020497 | 89.4kb | Anti-sense | Chr. 4 - 11704447 - 11714993 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm11832
Genomic Map