Assay Details
| Gene Symbol: Gm15816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282148.1 | NP_001269077.1 | 5 - 6 | 904 | 89 | |
| GenBank mRNA | AK042176.1 | - | 5 - 6 | 903 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | K(lysine) acetyltransferase 6A | Kat6a |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
-195.5kb | Anti-sense | Chr. 8 - 22859516 - 22943262 on NCBI Build GRCm38 |
| Downstream | microRNA 486 | Mir486 |
Mirn486, mir-486a, mmu-mir-486, mmu-mir-486a |
2.3kb | Anti-sense | Chr. 8 - 23142555 - 23142682 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15816
Genomic Map