Assay Details
| Gene Symbol: Orc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159563.1 | NP_001153035.1 | 12 - 13 | 1353 | 155 | |
| NM_015824.4 | NP_056639.3 | 12 - 13 | 1353 | 155 | ||
| GenBank mRNA | AB190257.2 | - | 12 - 13 | 1320 | 155 | |
| AJ132830.1 | - | 12 - 13 | 1335 | 155 | ||
| AK045388.1 | - | 12 - 13 | 1328 | 155 | ||
| AK086579.1 | - | 12 - 13 | 1356 | 155 | ||
| BC061252.1 | - | 12 - 13 | 1324 | 30250245 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Orc3
Genomic Map