Assay Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025669.1 | NP_079945.1 | 10 - 11 | 1898 | 123 | |
| GenBank mRNA | AK017507.1 | - | 10 - 11 | 1425 | 123 | |
| AK031964.1 | - | 10 - 11 | 1434 | 123 | ||
| BC060111.1 | - | 8 - 9 | 3180 | 123 | ||
| BC141031.1 | - | 10 - 11 | 1762 | 9055643 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnisr
Genomic Map