Assay Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025669.1 | NP_079945.1 | 4 - 5 | 1070 | 84 | |
| GenBank mRNA | AK012036.1 | - | 4 - 5 | 612 | 84 | |
| AK017507.1 | - | 4 - 5 | 597 | 84 | ||
| AK020035.1 | - | 4 - 5 | 598 | 84 | ||
| AK031964.1 | - | 4 - 5 | 606 | 84 | ||
| AK083021.1 | - | 4 - 5 | 596 | 84 | ||
| AK160288.1 | - | 4 - 5 | 596 | 84 | ||
| BC141031.1 | - | 4 - 5 | 934 | 9055643 | 84 | |
More Information
Set Membership: |
|

Pnisr
Genomic Map