Assay Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025669.1 | NP_079945.1 | 6 - 7 | 1431 | 66 | |
| GenBank mRNA | AK012036.1 | - | 6 - 7 | 973 | 66 | |
| AK017507.1 | - | 6 - 7 | 958 | 66 | ||
| AK020035.1 | - | 6 - 7 | 964 | 66 | ||
| AK031964.1 | - | 6 - 7 | 967 | 66 | ||
| AK077040.1 | - | 4 - 5 | 2667 | 66 | ||
| AK083021.1 | - | 6 - 7 | 957 | 66 | ||
| AK160288.1 | - | 6 - 7 | 957 | 66 | ||
| AK162695.1 | - | 6 - 7 | 926 | 66 | ||
| BC060111.1 | - | 6 - 7 | 944 | 66 | ||
| BC141031.1 | - | 6 - 7 | 1295 | 9055643 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022499 | AK031964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022499 | AK035634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022499 | NM_025669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119877 | BC060111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119877 | AK083021.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259768 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281626 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591944 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826282 | AK003061.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826282 | AK007944.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826282 | AK008678.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826282 | NM_029706.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167863 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107328 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131365 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145967 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119877 | BC060111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119877 | AK083021.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022499 | AK031964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022499 | AK035634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022499 | NM_025669.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05175510 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05175499 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107328 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131365 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145967 | - | - |
| Whole Mouse Genome G4122A | A_51_P277588 | AK031964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277588 | AK035634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P277588 | NM_025669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P80548 | BC141031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P80548 | BC060111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P80548 | AK017507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P80548 | AK031964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P80548 | NM_025669.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131365 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145967 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03424029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03436469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03519110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22775204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03656684 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10503571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503578 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503579 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503575 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435080_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453186_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437041_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429537_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453185_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429538_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435079_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503570 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511808 | - | - |
| Mouse Genome 430 2.0 Array | 1437041_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453185_at | - | - |
| Mouse Genome 430 2.0 Array | 1429538_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435079_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453186_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429537_at | - | - |
| Mouse Genome 430 2.0 Array | 1435079_at | - | - |
| Mouse Genome 430 2.0 Array | 1453185_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429538_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435080_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1453186_at | - | - |
| Mouse Genome 430 2.0 Array | 1437041_at | - | - |
| Mouse Genome 430 2.0 Array | 1435080_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429537_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66625_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66625_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400001214.mm.1 | NM_025669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400001214.mm.1 | NM_025669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400001214.mm.1 | TC0400000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400001213.mm.1 | NM_025669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400001213.mm.1 | NM_025669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400001213.mm.1 | TC0400000183.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435079_at | - | - |
| Mouse Expression Set 430 | 1453185_at | - | - |
| Mouse Expression Set 430 | 1429537_at | - | - |
| Mouse Expression Set 430 | 1435079_PM_at | - | - |
| Mouse Expression Set 430 | 1435080_x_at | - | - |
| Mouse Expression Set 430 | 1453186_at | - | - |
| Mouse Expression Set 430 | 1453186_PM_at | - | - |
| Mouse Expression Set 430 | 1435080_PM_x_at | - | - |
| Mouse Expression Set 430 | 1429538_a_at | - | - |
| Mouse Expression Set 430 | 1429537_PM_at | - | - |
| Mouse Expression Set 430 | 1453185_PM_at | - | - |
| Mouse Expression Set 430 | 1429538_PM_a_at | - | - |
| Mouse Expression Set 430 | 1437041_PM_at | - | - |
| Mouse Expression Set 430 | 1437041_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429537_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437041_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435080_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437041_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453185_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429537_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435079_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453186_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429538_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453185_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453186_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435080_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429538_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435079_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 568747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659283 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2614644 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659283 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2614644 | - | - |

Pnisr
Genomic Map