Assay Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025669.1 | NP_079945.1 | 7 - 8 | 1574 | 74 | |
| GenBank mRNA | AK012036.1 | - | 7 - 8 | 1116 | 74 | |
| AK017507.1 | - | 7 - 8 | 1101 | 74 | ||
| AK020035.1 | - | 7 - 8 | 1107 | 74 | ||
| AK031964.1 | - | 7 - 8 | 1110 | 74 | ||
| AK077040.1 | - | 5 - 6 | 2810 | 74 | ||
| AK083021.1 | - | 7 - 8 | 1100 | 74 | ||
| AK160288.1 | - | 7 - 8 | 1100 | 74 | ||
| BC060111.1 | - | 7 - 8 | 1087 | 74 | ||
| BC141031.1 | - | 7 - 8 | 1438 | 9055643 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnisr
Genomic Map