Assay Details
| Gene Symbol: Isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177663.4 | NP_808331.1 | 3 - 4 | 1441 | 73 | |
| GenBank mRNA | AK053778.1 | - | 3 - 4 | 1442 | 73 | |
| AK076747.1 | - | 3 - 4 | 1164 | 73 | ||
| AK155979.1 | - | 3 - 4 | 1445 | 73 | ||
| AK178428.1 | - | 1 - 2 | 75 | 73 | ||
| BC098326.1 | - | 3 - 4 | 1428 | 5720443 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310781 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310780 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P28792 | BC098326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P28792 | NM_177663.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P28792 | AK053778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P28792 | BC098326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P28792 | NM_177663.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P28792 | AK053778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028758 | BC098326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028758 | AK155979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028758 | NM_177663.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028758 | AK053778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934788 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 690551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10719 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178996 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140626 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054653 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028758 | BC098326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028758 | AK155979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028758 | NM_177663.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028758 | AK053778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P28792 | BC098326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P28792 | NM_177663.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P28792 | AK053778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P28792 | BC098326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P28792 | NM_177663.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P28792 | AK053778.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310795 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140626 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00693959 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178996 | - | - |
| Whole Mouse Genome G4122A | A_52_P28792 | BC098326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P28792 | NM_177663.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P28792 | AK053778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P28792 | BC098326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P28792 | NM_177663.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P28792 | AK053778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P572456 | BC098326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P572456 | AK076747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P572456 | AK155979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P572456 | AK178428.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P572456 | NM_177663.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101789 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178996 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02367975 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02367976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310780 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03061038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02930084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310796 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493104 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433960_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493103 | - | - |
| Mouse Genome 430 2.0 Array | 1433960_at | - | - |
| Mouse Genome 430 2.0 Array | 1433960_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229504_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229504_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300004790.mm.1 | NM_177663.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004790.mm.1 | NM_177663.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004790.mm.1 | TC0300000709.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004789.mm.1 | NM_177663.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004789.mm.1 | NM_177663.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004789.mm.1 | TC0300000709.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433960_at | - | - |
| Mouse Expression Set 430 | 1433960_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433960_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433960_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 872493 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226070 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226070 | - | - |
| mouse-6 v1.1 (Illumina) | scl23014.5.1_141-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23014.5.1_141-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2996667 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2996667 | - | - |

Isg20l2
Genomic Map