Assay Details
| Gene Symbol: Orc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159563.1 | NP_001153035.1 | 16 - 17 | 1737 | 85 | |
| NM_015824.4 | NP_056639.3 | 16 - 17 | 1740 | 85 | ||
| GenBank mRNA | AB190257.2 | - | 16 - 17 | 1704 | 85 | |
| AJ132830.1 | - | 16 - 17 | 1722 | 85 | ||
| AK045388.1 | - | 16 - 17 | 1715 | 85 | ||
| AK078115.1 | - | 11 - 12 | 1251 | 85 | ||
| AK086579.1 | - | 16 - 17 | 1743 | 85 | ||
| AK145324.1 | - | 16 - 17 | 1719 | 85 | ||
| BC002204.1 | - | 1 - 2 | 286 | 85 | ||
| BC061252.1 | - | 16 - 17 | 1708 | 30250245 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Orc3
Genomic Map