Assay Details
| Gene Symbol: Pgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175013.2 | NP_778178.3 | 8 - 9 | 1352 | 88 | |
| GenBank mRNA | AK035507.1 | - | 8 - 9 | 1352 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgm5
Genomic Map