Assay Details
| Gene Symbol: Stat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011486.4 | NP_035616.1 | 5 - 6 | 741 | 69 | |
| NM_213659.2 | NP_998824.1 | 5 - 6 | 741 | 69 | ||
| NM_213660.2 | NP_998825.1 | 5 - 6 | 741 | 69 | ||
| GenBank mRNA | AK145486.1 | - | 5 - 6 | 597 | 69 | |
| AK153005.1 | - | 5 - 6 | 653 | 69 | ||
| AK153472.1 | - | 5 - 6 | 653 | 69 | ||
| AY299489.1 | - | 4 - 5 | 468 | 69 | ||
| AY299490.1 | - | 4 - 5 | 468 | 69 | ||
| BC003806.1 | - | 5 - 6 | 629 | 3593588 | 69 | |
| BC019168.1 | - | 5 - 6 | 645 | 4923137 | 69 | |
| L29278.1 | - | 5 - 6 | 720 | 69 | ||
| U06922.1 | - | 5 - 6 | 530 | 69 | ||
| U08378.1 | - | 5 - 6 | 608 | 69 | ||
| U30709.1 | - | 5 - 6 | 634 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stat3
Genomic Map