Assay Details
| Gene Symbol: Sim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011377.2 | NP_035507.2 | 3 - 4 | 906 | 82 | |
| GenBank mRNA | D63383.1 | - | 3 - 4 | 912 | 82 | |
| D64135.1 | - | 3 - 4 | 853 | 82 | ||
| U40576.1 | - | 3 - 4 | 1238 | 82 | ||
| U42554.1 | - | 3 - 4 | 578 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sim2
Genomic Map