Assay Details
| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293741.1 | NP_001280670.1 | 3 - 4 | 232 | 145 | |
| NM_026677.4 | NP_080953.1 | 3 - 4 | 363 | 145 | ||
| GenBank mRNA | AB232608.1 | - | 3 - 4 | 248 | 145 | |
| AK002303.1 | - | 3 - 4 | 330 | 145 | ||
| AK080805.1 | - | 3 - 4 | 233 | 145 | ||
| BB867138.1 | - | 3 - 4 | 366 | 145 | ||
| BC027214.1 | - | 3 - 4 | 356 | 3709140 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293741.1 transcripts.
Set Membership: |
|

Rab13
Genomic Map