Assay Details
| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293741.1 | NP_001280670.1 | 6 - 7 | 466 | 76 | |
| NM_026677.4 | NP_080953.1 | 6 - 7 | 597 | 76 | ||
| GenBank mRNA | AB232608.1 | - | 6 - 7 | 482 | 76 | |
| AK002303.1 | - | 6 - 7 | 564 | 76 | ||
| AK080805.1 | - | 6 - 7 | 467 | 76 | ||
| BC027214.1 | - | 6 - 7 | 590 | 3709140 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab13
Genomic Map