| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900009723.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009723.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009723.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009723.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009723.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009722.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009722.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009722.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009722.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009722.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009721.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009721.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009721.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009721.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009721.mm.1 |
TC1900001039.mm.1 |
MTA TC |