Assay Details
| Gene Symbol: Mark2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080388.2 | NP_001073857.1 | 8 - 9 | 1407 | 85 | |
| NM_001080389.2 | NP_001073858.1 | 8 - 9 | 1407 | 85 | ||
| NM_001080390.2 | NP_001073859.1 | 8 - 9 | 1124 | 85 | ||
| NM_007928.3 | NP_031954.2 | 8 - 9 | 1407 | 85 | ||
| GenBank mRNA | AK045329.1 | - | 8 - 9 | 981 | 85 | |
| AK154864.1 | - | 8 - 9 | 1124 | 85 | ||
| AK172444.1 | - | 8 - 9 | 1367 | 85 | ||
| AK172664.1 | - | 8 - 9 | 1050 | 85 | ||
| AK220191.1 | - | 9 - 10 | 973 | 85 | ||
| BC058556.1 | - | 8 - 9 | 1344 | 5289799 | 85 | |
| X70764.1 | - | 8 - 9 | 861 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mark2
Genomic Map