Assay Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024434.6 | NP_077754.3 | 10 - 11 | 1265 | 121 | |
| GenBank mRNA | AF334160.1 | - | 10 - 11 | 1226 | 121 | |
| AK002819.1 | - | 10 - 11 | 1201 | 121 | ||
| AK005334.1 | - | 10 - 11 | 1178 | 121 | ||
| AK008600.1 | - | 10 - 11 | 1152 | 121 | ||
| AK010384.1 | - | 10 - 11 | 1149 | 121 | ||
| AK010502.1 | - | 10 - 11 | 1218 | 121 | ||
| AK011604.1 | - | 10 - 11 | 1151 | 121 | ||
| AK166958.1 | - | 10 - 11 | 1149 | 121 | ||
| AK169032.1 | - | 10 - 11 | 1151 | 121 | ||
| AK185578.1 | - | 2 - 3 | 162 | 121 | ||
| BC016536.1 | - | 10 - 11 | 1265 | 3488469 | 121 | |
| CT010237.1 | - | 10 - 11 | 1179 | 121 | ||
| CT010301.1 | - | 10 - 11 | 1179 | 121 | ||
More Information
Set Membership: |
|

Lap3
Genomic Map