Assay Details
| Gene Symbol: Chrna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015730.5 | NP_056545.3 | 2 - 3 | 335 | 69 | |
| GenBank mRNA | AF225912.2 | - | 2 - 3 | 233 | 69 | |
| AK034228.1 | - | 2 - 3 | 346 | 69 | ||
| AK083157.1 | - | 2 - 3 | 266 | 69 | ||
| AK136886.1 | - | 2 - 3 | 322 | 69 | ||
| AK143938.1 | - | 2 - 3 | 343 | 69 | ||
| AY574263.1 | - | 2 - 3 | 231 | 69 | ||
| BC053013.1 | - | 2 - 3 | 335 | 5706867 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chrna4
Genomic Map