Assay Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147201.2 | NP_671734.1 | 2 - 3 | 320 | 62 | |
| GenBank mRNA | AF302138.1 | - | 2 - 3 | 319 | 62 | |
| AK046142.1 | - | 2 - 3 | 314 | 62 | ||
| AK077440.1 | - | 2 - 3 | 356 | 62 | ||
| AK170501.1 | - | 2 - 3 | 336 | 62 | ||
| AK191367.1 | - | 3 - 4 | 200 | 62 | ||
| AK217229.1 | - | 1 - 2 | 247 | 62 | ||
| BC004756.1 | - | 2 - 3 | 290 | 3154089 | 62 | |
| BC018463.1 | - | 2 - 3 | 294 | 3598438 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrbp1
Genomic Map