Assay Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147201.2 | NP_671734.1 | 5 - 6 | 631 | 106 | |
| GenBank mRNA | AF302138.1 | - | 5 - 6 | 630 | 106 | |
| AK046142.1 | - | 5 - 6 | 625 | 106 | ||
| AK077440.1 | - | 5 - 6 | 667 | 106 | ||
| AK170501.1 | - | 5 - 6 | 647 | 106 | ||
| BC004756.1 | - | 5 - 6 | 601 | 3154089 | 106 | |
| BC018463.1 | - | 5 - 6 | 605 | 3598438 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrbp1
Genomic Map