Assay Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147201.2 | NP_671734.1 | 9 - 10 | 909 | 71 | |
| GenBank mRNA | AF302138.1 | - | 9 - 10 | 908 | 71 | |
| AK046142.1 | - | 10 - 11 | 927 | 71 | ||
| AK077440.1 | - | 9 - 10 | 945 | 71 | ||
| AK170501.1 | - | 9 - 10 | 925 | 71 | ||
| BC004756.1 | - | 9 - 10 | 879 | 3154089 | 71 | |
| BC018463.1 | - | 9 - 10 | 883 | 3598438 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrbp1
Genomic Map