Assay Details
| Gene Symbol: Coq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172687.1 | NP_766275.1 | 2 - 3 | 255 | 89 | |
| GenBank mRNA | AK028680.1 | - | 2 - 3 | 255 | 89 | |
| AK035318.1 | - | 2 - 3 | 250 | 89 | ||
| AK142287.1 | - | 2 - 3 | 193 | 89 | ||
| AK166475.1 | - | 2 - 3 | 210 | 89 | ||
| AK170488.1 | - | 2 - 3 | 250 | 89 | ||
| BC060960.1 | - | 2 - 3 | 257 | 30312962 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coq3
Genomic Map