Assay Details
| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011881.3 | NP_036011.3 | 3 - 4 | 1022 | 64 | |
| GenBank mRNA | BC075722.1 | - | 2 - 3 | 2346 | 64 | |
| BC096611.1 | - | 3 - 4 | 1022 | 5357325 | 64 | |
More Information
Set Membership: |
|

Grk1
Genomic Map