Assay Details
| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011881.3 | NP_036011.3 | 5 - 6 | 1231 | 129 | |
| GenBank mRNA | BC075722.1 | - | 4 - 5 | 2555 | 129 | |
| BC096611.1 | - | 5 - 6 | 1231 | 5357325 | 129 | |
More Information
Set Membership: |
|

Grk1
Genomic Map