Assay Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003508.1 | 10 - 11 | 1372 | 95 | ||
| GenBank mRNA | AK143831.1 | - | 10 - 11 | 1373 | 95 | |
| AK147949.1 | - | 1992 | 95 | |||
| J03368.1 | - | 10 - 11 | 1370 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 3, member B |
Fam3b |
2-21, 9030624C24Rik, D16Jhu19e, ORF9, Pander |
-47.0kb | Anti-sense | Chr. 16 - 97471086 - 97504936 on NCBI Build GRCm38 |
| Downstream |
transmembrane protease, serine 2 |
Tmprss2 | D16Ertd61e | 8.6kb | Anti-sense | Chr. 16 - 97564682 - 97659538 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mx2
Genomic Map