Assay Details
| Gene Symbol: Ubxn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026534.2 | NP_080810.2 | 3 - 4 | 365 | 127 | |
| GenBank mRNA | AB120715.1 | - | 3 - 4 | 358 | 127 | |
| AK012822.1 | - | 3 - 4 | 364 | 127 | ||
| AK013985.1 | - | 3 - 4 | 360 | 127 | ||
| AK028268.1 | - | 3 - 4 | 368 | 127 | ||
| AK032332.1 | - | 3 - 4 | 365 | 127 | ||
| AK049018.1 | - | 3 - 4 | 373 | 127 | ||
| BC076632.1 | - | 3 - 4 | 365 | 30637967 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn2b
Genomic Map