Assay Details
| Gene Symbol: Spaca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024279.2 | NP_077241.1 | 1 - 2 | 154 | 63 | |
| GenBank mRNA | AK018926.1 | - | 1 - 2 | 155 | 63 | |
| BC048522.1 | - | 1 - 2 | 154 | 6741699 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spaca7
Genomic Map