Assay Details
| Gene Symbol: Spaca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024279.2 | NP_077241.1 | 6 - 7 | 591 | 111 | |
| GenBank mRNA | AK018926.1 | - | 6 - 7 | 592 | 111 | |
| BC048522.1 | - | 6 - 7 | 587 | 6741699 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275053 | BC048522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275053 | NM_024279.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275053 | AK018926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275053 | BC048522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275053 | NM_024279.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275053 | AK018926.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 792210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771649 | BC026638.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771649 | BC031613.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64032 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132216 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155332 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275053 | BC048522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275053 | NM_024279.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275053 | AK018926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275053 | BC048522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275053 | NM_024279.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275053 | AK018926.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25662412 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06427422 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132216 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155332 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01676936 | - | - |
| Whole Mouse Genome G4122A | A_51_P275053 | BC048522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275053 | NM_024279.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275053 | AK018926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275053 | BC048522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275053 | NM_024279.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275053 | AK018926.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129466 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130687 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155332 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132216 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318465 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318458 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318468 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318463 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07318473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25662391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06738070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06726869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06695475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07338657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06729793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07266035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07191675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07191685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06729795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07149646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07124409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07149650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07191690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07225831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07119867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06726873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07318457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25662431 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570197 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570196 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429757_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570191 | - | - |
| Mouse Genome 430 2.0 Array | 1429757_at | - | - |
| Mouse Genome 430 2.0 Array | 1429757_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78634_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78634_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800000884.mm.1 | NM_024279.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800000884.mm.1 | NM_024279.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800000884.mm.1 | TC0800000108.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429757_at | - | - |
| Mouse Expression Set 430 | 1429757_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429757_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429757_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 550728 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1190154 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242313 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242313 | - | - |
| mouse-6 v1.1 (Illumina) | scl34069.7.1_94-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190154 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34069.7.1_94-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190154 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242313 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242313 | - | - |

Spaca7
Genomic Map