Assay Details
| Gene Symbol: Adgrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014394.2 | NP_001014416.2 | 9 - 10 | 1424 | 75 | |
| GenBank mRNA | AY701869.1 | - | 9 - 10 | 1424 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058947 | NM_001014394.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058947 | AY701869.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139905 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163515 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112577 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058947 | NM_001014394.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058947 | AY701869.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23674 | NM_001014394.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23674 | AY255593.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23674 | AY701869.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23571360 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163515 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112577 | - | - |
| Whole Mouse Genome G4122A | A_52_P23674 | NM_001014394.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P23674 | AY255593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P23674 | AY701869.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163515 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23571342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04296358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04715759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04947443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04464849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04380175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04947437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04715758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04715757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04638033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04877398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04938079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04873613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04638035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04534512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04406613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04579539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04743805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04157263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04638032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04773093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23571373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04579543 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10528957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528968 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528965 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528966 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528956 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528951 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500019558.mm.1 | TC0500002155.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019558.mm.1 | NM_001014394.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019558.mm.1 | NM_001014394.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019557.mm.1 | TC0500002155.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019557.mm.1 | NM_001014394.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019557.mm.1 | NM_001014394.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 767448 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2537217 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2529810 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2909568 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2909568 | - | - |

Adgrf3
Genomic Map