Assay Details
| Gene Symbol: Psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289476.1 | NP_001276405.1 | 4 - 5 | 1206 | 70 | |
| NM_011969.2 | NP_036099.1 | 4 - 5 | 567 | 70 | ||
| GenBank mRNA | AF019662.1 | - | 4 - 5 | 505 | 70 | |
| AK088615.1 | - | 4 - 5 | 512 | 70 | ||
| AK146765.1 | - | 4 - 5 | 524 | 70 | ||
| AK165563.1 | - | 4 - 5 | 520 | 70 | ||
| AK168234.1 | - | 4 - 5 | 538 | 70 | ||
| BC008222.1 | - | 4 - 5 | 500 | 2581897 | 70 | |
| BC014752.1 | - | 4 - 5 | 1154 | 70 | ||
| DV049235.1 | - | 4 - 5 | 536 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psma7
Genomic Map