Assay Details
| Gene Symbol: Psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289476.1 | NP_001276405.1 | 6 - 7 | 138 | ||
| NM_011969.2 | NP_036099.1 | 6 - 7 | 138 | |||
| GenBank mRNA | AF019662.1 | - | 6 - 7 | 138 | ||
| AK088615.1 | - | 6 - 7 | 696 | 138 | ||
| AK146765.1 | - | 6 - 7 | 138 | |||
| AK165563.1 | - | 6 - 7 | 138 | |||
| AK168234.1 | - | 6 - 7 | 722 | 138 | ||
| BC008222.1 | - | 6 - 7 | 684 | 2581897 | 138 | |
| BC014752.1 | - | 6 - 7 | 1338 | 138 | ||
| DV049235.1 | - | 6 - 7 | 138 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Psma7
Genomic Map