Assay Details
| Gene Symbol: Nkain4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021426.4 | NP_067401.3 | 5 - 6 | 604 | 62 | |
| GenBank mRNA | AB030182.1 | - | 5 - 6 | 582 | 62 | |
| AK005262.1 | - | 5 - 6 | 575 | 62 | ||
| AK007632.1 | - | 5 - 6 | 569 | 62 | ||
| BC061107.1 | - | 5 - 6 | 589 | 30295742 | 62 | |
| BC089330.1 | - | 5 - 6 | 556 | 30358178 | 62 | |
| EF058055.1 | - | 5 - 6 | 529 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nkain4
Genomic Map