Assay Details
| Gene Symbol: Psapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175249.3 | NP_780458.2 | 1 | 2421 | 140 | |
| GenBank mRNA | AV088427.2 | - | 1 | 932 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AK028455.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AK078699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AV088427.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | NM_175249.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AK009408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AK028455.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AK078699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AV088427.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | NM_175249.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P523368 | AK009408.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122442 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AK028455.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AK078699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AV088427.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | NM_175249.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AK009408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AK028455.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AK078699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AV088427.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | NM_175249.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P523368 | AK009408.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23605990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23606008 | - | - |
| Whole Mouse Genome G4122A | A_51_P345073 | AK028455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345073 | AK078699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345073 | AV088427.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345073 | NM_175249.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345073 | AK009408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AK028455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AK078699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AV088427.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | NM_175249.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AK009408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AK028455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AK078699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AV088427.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | NM_175249.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P523368 | AK009408.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105207 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928862 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928853 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928873 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928885 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928881 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23606003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23605972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04848531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04691833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04311885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04279755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04622510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23606023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23605969 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10521460 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429457_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440186_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10521459 | - | - |
| Mouse Genome 430 2.0 Array | 1440186_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429457_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429457_at | - | - |
| Mouse Genome 430 2.0 Array | 1440186_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76943_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76943_PM_at | - | - |
| Mouse Expression Set 430 | 1429457_PM_at | - | - |
| Mouse Expression Set 430 | 1429457_at | - | - |
| Mouse Expression Set 430 | 1440186_PM_s_at | - | - |
| Mouse Expression Set 430 | 1440186_s_at | - | - |
| Murine Genome U74v2 Set | 100043_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100043_f_at | - | - |
| Murine Genome U74v2 Set | 168086_r_at | - | - |
| Murine Genome U74v2 Set | 168086_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429457_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429457_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429457_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429457_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440186_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440186_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 573265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3520735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003520735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635197 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2635197 | - | - |
| mouse-6 v1.1 (Illumina) | scl27892.1.2_292-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3520735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27892.1.2_292-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2635197 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2635197 | - | - |

Psapl1
Genomic Map