Assay Details
| Gene Symbol: 4930425K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038182.1 | 1 - 2 | 160 | 117 | ||
| GenBank mRNA | AK015200.1 | - | 1 - 2 | 162 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, aminophospholipid transporter (APLT), class I, type 8A, member 1 |
Atp8a1 |
AI481521, AI853962, APLT, AW743152, AW822227, Atp3a2, B230107D19Rik, ClassI |
-91.0kb | Sense | Chr. 5 - 67618139 - 67847487 on NCBI Build GRCm38 |
| Downstream |
glutaredoxin, cysteine rich 1 |
Grxcr1 |
Tg(Eno2-Gabrb3)0370Brll, pi, tde |
88.8kb | Anti-sense | Chr. 5 - 68031835 - 68166398 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930425K10Rik
Genomic Map