Assay Details
| Gene Symbol: Gm5108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256184.1 | NP_001243113.1 | 2 - 3 | 274 | 60 | |
| GenBank mRNA | AK076825.1 | - | 2 - 3 | 276 | 60 | |
| BC120772.1 | - | 2 - 3 | 204 | 60 | ||
| BC120798.1 | - | 2 - 3 | 204 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930425K10 gene | 4930425K10Rik | -1.5kb | Anti-sense | Chr. 5 - 67933612 - 67943081 on NCBI Build GRCm38 | |
| Downstream |
glutaredoxin, cysteine rich 1 |
Grxcr1 |
Tg(Eno2-Gabrb3)0370Brll, pi, tde |
56.7kb | Sense | Chr. 5 - 68031835 - 68166398 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5108
Genomic Map