Assay Details
| Gene Symbol: Mrps31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020560.2 | NP_065585.1 | 4 - 5 | 827 | 112 | |
| GenBank mRNA | AK005110.1 | - | 4 - 5 | 786 | 112 | |
| AK011825.1 | - | 4 - 5 | 783 | 112 | ||
| AK078073.1 | - | 4 - 5 | 770 | 112 | ||
| AK161913.1 | - | 4 - 5 | 770 | 112 | ||
| BC006764.1 | - | 4 - 5 | 720 | 112 | ||
| BC027430.1 | - | 4 - 5 | 731 | 4187438 | 112 | |
| Z46966.1 | - | 4 - 5 | 746 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps31
Genomic Map