Assay Details
| Gene Symbol: Ccser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027045.1 | NP_081321.1 | 2 - 3 | 1673 | 105 | |
| GenBank mRNA | AK013689.1 | - | 2 - 3 | 1627 | 105 | |
| AK147383.1 | - | 2 - 3 | 1668 | 105 | ||
| AK172153.1 | - | 2 - 3 | 1647 | 105 | ||
| BC111465.1 | - | 1 - 2 | 54 | 105 | ||
| BC139090.1 | - | 1 - 2 | 1461 | 8862112 | 105 | |
More Information
Set Membership: |
|

Ccser2
Genomic Map